Order Entry
Puerto Rico
ContactUsLinkComponent
Human RUNX2 ELISA Kit
Human RUNX2 ELISA Kit
Catalog # MSPP-SEB011HU
Supplier:  Cloud-Clone
CAS Number:  
undefined
Human RUNX2 ELISA Kit
Catalog # MSPP-SEB011HU
Supplier:  Cloud-Clone
Supplier Number:  SEB011HU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Human
  • Target protein:
    RUNX2
  • Size:
    1 kit
  • Sample Type:
    Tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Runt Related Transcription Factor 2 (RUNX2) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    0.156 - 10 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    0.054 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEB011HU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Human RUNX2 (Runt Related Transcription Factor 2). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.054 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

Cbfa1/Runx2 is a key transcription factor associated with osteoblast differentiation.This protein is a member of the RUNX family of transcription factors and has a Runt DNA-binding domain. It is essential for osteoblastic differentiation and skeletal morphogenesis and acts as a scaffold for nucleic acids and regulatory factors involved in skeletal gene expression. The protein can bind DNA both as a monomer or, with more affinity, as a subunit of a heterodimeric complex. Transcript variants of the gene that encode different protein isoforms result from the use of alternate promoters as well as alternate splicing.The RUNX2 transcription factor is the master regulator of osteoblast differentiation is required for chondrocyte hypertrophy.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.