Order Entry
Puerto Rico
ContactUsLinkComponent
Human PSAP ELISA Kit
Human PSAP ELISA Kit
Catalog # MSPP-SEC756HU
CAS Number:  
undefined
Human PSAP ELISA Kit
Catalog # MSPP-SEC756HU
Supplier Number:  SEC756HU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Human
  • Target protein:
    PSAP
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cerebrospinal fluid, seminal plasma, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Prosaposin (PSAP) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    0.78 - 50 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    0.33 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEC756HU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Human PSAP (Prosaposin). The assay range is from 0.78 to 50 ng/ml (Sandwich kit) with a sensitivity of 0.33 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

Prosaposin also known as PSAP is a highly conserved glycoprotein which is a precursor for four cleavage products: saposins A, B, C, and D. Saposin is an acronym for Sphingolipid Activator PrO[S]teINs. Each domain of the precursor protein is approximately 80 amino acid residues long with nearly identical placement of cysteine residues and glycosylation sites. Saposins A-D localize primarily to the lysosomal compartment where they facilitate the catabolism of glycosphingolipids with short oligosaccharide groups. The precursor protein exists both as a secretory protein and as an integral membrane protein and has neurotrophic activities. Saposins A-D are required for the hydrolysis of certain shingolipids by specific lysosomal hydrolases.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.