- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Conjugate ELISA:Biotin
- Format:Pre-coated
- Host:Rabbit
- Primary antibody reactivity:Human
- Target protein:PIICP
- Size:1 kit
- Sample Type:Serum, plasma and other biological fluids
- Cross Reactivity:No significant cross-reactivity or interference between Procollagen II C-Terminal Propeptide (PIICP) and analogues was observed
- Detection Method:Colorimetric
- Time to Results:3 h
- Shelf Life:12 Months
- Detection Range:125 - 2000 pg/ml
- Storage Temperature:4 °C for one month (frequent use), −20 °C for one year
- Sample Volume:100 µl
- Sensitivity:57 pg/ml
- Regulatory Status:RUO
- Cat. No.:MSPP-SEA964HU
- No. of tests:96 wells
This assay has high sensitivity and excellent specificity for detecting Human PIICP (Procollagen II C-Terminal Propeptide). The assay range is from 125 to 2000 pg/ml (Sandwich kit) with a sensitivity of 57 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
- High sensitivity and specificity
- Perfect reproducibility and consistency across batches
- Quality control with three-level inspections
- Wide range of targets/species available
- Intra-assay: CV<10%; Inter-assay: CV<12%
Procollagen II C peptide is used as a joint fluid marker of collagen synthesis by chondrocytes in patients with osteoarthritis. Since osteochondromatosis activates ectopic chondrogenesis in the synovial membrane, procollagen II C peptide is expected to be a good marker of the disease. Procollagen II C peptide can be used to evaluate the condition of the disease metabolically by simply aspirating the joint fluid.the concentration of procollagen II C peptide is a good marker of chondrogenesis by osteochondromatosis. Body mass index showed a slightly positive association with baseline PIICP level. In multiple linear regression analyses adjusted for age and body mass index, radiographic JSN over four years had a direct positive correlation with baseline PIICP leve.the prognostic value of procollagen type II carboxy-terminal propeptide (PIICP) level in synovial fluid in relation to early tibiofemoral joint osteoarthritis (OA).