Order Entry
Puerto Rico
ContactUsLinkComponent
Human OPN ELISA Kit
Human OPN ELISA Kit
Catalog # MSPP-SEA899HU
CAS Number:  
undefined
Human OPN ELISA Kit
Catalog # MSPP-SEA899HU
Supplier Number:  SEA899HU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Human
  • Target protein:
    OPN
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Osteopontin (OPN) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    0.625 - 40 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    0.237 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEA899HU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Human OPN (Osteopontin). The assay range is from 0.625 to 40 ng/ml (Sandwich kit) with a sensitivity of 0.237 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

Osteopontin (OPN), is a secreted multifunctional glyco-phosphoprotein with roles in bone metabolism, immune regulation, tissue remodeling , cell survival, and tumor progression. Gene structure and chromosomal location identify OPN as a member of the small integrin-binding ligand N-linked glycoprotein (SIBLING) family that also includes BSP,BMP1,DSPP, ENAM and MEPE. Murine OPN issynthesized as a 294 amino acid (aa) precursor protein with a predicted 16 aa signal peptide and a highly unusual mature protein sequence, containing 68 acidic aa and 23 potential Ser/Thr phosphorylation sites. Although the predicted molecular weight of OPN is 31 kDa,phosphorylation and N- and O-glycosylation may allow it to appear as large as 75 kDa. Variability in post-translational modifications can influence the activity of OPN.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.