Order Entry
Puerto Rico
ContactUsLinkComponent
Human mTOR ELISA Kit
Human mTOR ELISA Kit
Catalog # MSPP-SEB806HU
CAS Number:  
undefined
Human mTOR ELISA Kit
Catalog # MSPP-SEB806HU
Supplier Number:  SEB806HU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Human
  • Target protein:
    mTOR
  • Size:
    1 kit
  • Sample Type:
    Tissue homogenates, cell lysates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Serine/threonine-protein kinase mTOR (mTOR) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    0.312 - 20 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    0.119 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEB806HU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Human mTOR (Serine/threonine-protein kinase mTOR). The assay range is from 0.312 to 20 ng/ml (Sandwich kit) with a sensitivity of 0.119 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

The mammalian target of rapamycin (mTOR) also known as FK506 binding protein 12-rapamycin associated protein 1 (FRAP1) is a protein which in humans is encoded by the FRAP1 gene.mTOR is a serine/threonine protein kinase that regulates cell growth, cell proliferation, cell motility, cell survival, protein synthesis, and transcription. Current research indicates that mTOR integrates the input from multiple upstream pathways, including insulin, growth factors (such as IGF-1 and IGF-2), and mitogens. mTOR also functions as a sensor of cellular nutrient and energy levels and redox status. The disregulation of the mTOR pathway is implicated as a contributing factor to various human disease processes, especially various types of cancer.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.