Order Entry
Puerto Rico
ContactUsLinkComponent
Human LCAT ELISA Kit
Human LCAT ELISA Kit
Catalog # MSPP-SEJ516HU
CAS Number:  
undefined
Human LCAT ELISA Kit
Catalog # MSPP-SEJ516HU
Supplier Number:  SEJ516HU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Human
  • Target protein:
    LCAT
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Lecithin Cholesterol Acyltransferase (LCAT) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    0.94 - 60 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    0.41 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEJ516HU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Human LCAT (Lecithin Cholesterol Acyltransferase). The assay range is from 0.94 to 60 ng/ml (Sandwich kit) with a sensitivity of 0.41 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

Lecithin-cholesterol acyltransferase is an enzyme that converts free cholesterol into cholesteryl ester (a more hydrophobic form of cholesterol), which is then sequestered into the core of a lipoprotein particle, eventually making the newly synthesized HDL spherical and forcing the reaction to become unidirectional since the particles are removed from the surface. The enzyme is bound to high-density lipoproteins (HDLs) and low-density lipoproteins in the blood plasma. An unusual feature of the message is that the poly(A) signal appears to overlap the COOH-terminal glutamic acid and stop codons. The protein has several extended sequences of hydrophobic amino acids, one of which is similar to sequences in pancreatic lipase and lingual lipase.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.