Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human KNG1 ELISA Kit
Catalog # MSPP-SEB267HU
CAS Number:  
undefined
Human KNG1 ELISA Kit
Catalog # MSPP-SEB267HU
Supplier Number:  SEB267HU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Human
  • Target protein:
    KNG1
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Kininogen 1 (KNG1) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    0.156 - 10 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    0.063 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEB267HU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Human KNG1 (Kininogen 1). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.063 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

High molecular weight kininogen (HMWK) plays an important role in assembly of the plasma kallikrein -kinin system. The KNG1 gene generates both HMWK and low molecular weight kininogen (LMWK) through alternative splicing. Both HMWK and LMWK contain an identical heavy chain consisting of protein domains 1, 2, and 3. However, HMWK contains a 56 kD light chain that consists of domains 5 and 6H, whereas LMWK contains a unique 4 kD light chain that consists of domain 5 L. In both proteins, the heavy and light chains are linked by domain 4, which contains the bradykinin (BK) nonapeptide. BK, which is released by plasma kallikrein, is a potent inflammatory mediator that causes vasodilation and enhanced capillary permeability, induces pain, and stimulates production of nitric oxide and prostacyclin from endothelial cells.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.