Order Entry
Puerto Rico
ContactUsLinkComponent
Human IL25 ELISA Kit
Human IL25 ELISA Kit
Catalog # MSPP-SEB694HU
CAS Number:  
undefined
Human IL25 ELISA Kit
Catalog # MSPP-SEB694HU
Supplier Number:  SEB694HU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Human
  • Target protein:
    IL25
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Interleukin 25 (IL25) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    15.6 - 1000 pg/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    6.4 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEB694HU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Human IL25 (Interleukin 25). The assay range is from 15.6 to 1000 pg/ml (Sandwich kit) with a sensitivity of 6.4 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

Interleukin-25 (IL-25) is a cytokine that belongs to the IL-17 cytokine family and is secreted by type 2 helper T cells (Th2) and mast cells. It is also known as IL-17E. IL-25 induces the production of other cytokines, including IL-4, IL-5 and IL-13 in multiple tissues, which stimulate the expansion of eosinophils. This cytokine is an important molecule controlling immunity of the gut and has been implicated in chronic inflammation associated with the gastrointestinal tract. Further, the IL-25 gene has been identified in a chromosomal region associated with autoimmune diseases of the gut such as inflammatory bowel disease (IBD), although no direct evidence suggests that IL-25 plays any role in this disease.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.