Order Entry
Puerto Rico
ContactUsLinkComponent
Human IL22 ELISA Kit
Human IL22 ELISA Kit
Catalog # MSPP-SEC032HU
CAS Number:  
undefined
Human IL22 ELISA Kit
Catalog # MSPP-SEC032HU
Supplier Number:  SEC032HU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Human
  • Target protein:
    IL22
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Interleukin 22 (IL22) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    15.6 - 1000 pg/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    5.7 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEC032HU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Human IL22 (Interleukin 22). The assay range is from 15.6 to 1000 pg/ml (Sandwich kit) with a sensitivity of 5.7 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

IL-22, a class of potent mediators of cellular inflammatory responses. It shares us of IL-10R2 in cell signaling with other members of this family, IL-10, IL-26, IL-28, IL-29. IL-22 is produced by activated DC and T cells and initiates innate immune responses against bacterial pathogens especially in epithelial cells such as respiratory and gut epithelial cells. IL-22 along with IL-17 is rapidly produced by splenic LTi-like cells  and can be also produced by Th17 cells and likely plays a role in the coordinated response of both adaptive and innate immune systems. IL-22 biological activity is initiated by binding to a cell-surface complex composed of IL-22R1 and IL-10R2 receptor chains and further regulated by interactions with a soluble binding protein, IL-22BP, which shares sequence similarity with an extracellular region of IL-22R1.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.