Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Human GPX1 ELISA Kit

Human GPX1 ELISA Kit

96 wells, 1 kit
Catalog # MSPP-SEA295HU
Marketsource
Supplier:   Cloud-Clone
Supplier #:  SEA295HU
Human GPX1 ELISA Kit
Catalog # MSPP-SEA295HU
Supplier:  Cloud-Clone
Marketsource
CAS Number:  
Human GPX1 ELISA Kit
Human GPX1 ELISA Kit

96 wells, 1 kit

CAS Number:  
Price
$630.00
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

This assay has high sensitivity and excellent specificity for detecting Human GPX1 (Glutathione Peroxidase 1). The assay range is from 3.12 to 200 ng/ml (Sandwich kit) with a sensitivity of 1.25 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

Glutathione peroxidase is the general name of an enzyme family with peroxidase activity whose main biological role is to protect the organism from oxidative damage. The biochemical function of glutathione peroxidase is to reduce lipid hydroperoxides to their corresponding alcohols and to reduce free hydrogen peroxide to water. There are several isozymes encoded by different genes, which vary in celullar location and substrate specificity. Glutathione peroxidase 1 is the most abundant version, found in the cytoplasm of nearly all mammalian tissues, whose preferred substrate is hydrogen peroxide. Glutathione peroxidase is a selenium-containing tetrameric glycoprotein, that is, a molecule with four selenocysteine amino acid residues.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Documents

Specifications

Specifications

  • Size:
    1 kit
  • No. of tests:
    96 wells
  • Primary antibody reactivity:
    Human
  • Detection Range:
    3.12 - 200 ng/ml
  • Sample Volume:
    100 µl
  • Host:
    Rabbit
  • Time to Results:
    3 h
  • Detection Method:
    Colorimetric
  • Cat. No.:
    MSPP-SEA295HU
  • Regulatory Status:
    RUO
  • Assay duration:
    Multiple steps
  • Sensitivity:
    1.25 ng/ml
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Assay Type (ELISA with LOV):
    Sandwich
  • Target protein:
    GPX1
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Shelf Life:
    12 Months
  • Cross Reactivity:
    No significant cross-reactivity or interference between Glutathione Peroxidase 1 (GPX1) and analogues was observed
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year

Specifications