Order Entry
Puerto Rico
ContactUsLinkComponent
Human GFAP ELISA Kit
Human GFAP ELISA Kit
Catalog # MSPP-SEA068HU
CAS Number:  
undefined
Human GFAP ELISA Kit
Catalog # MSPP-SEA068HU
Supplier Number:  SEA068HU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Human
  • Target protein:
    GFAP
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Glial Fibrillary Acidic Protein (GFAP) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    0.156 - 10 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    0.056 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEA068HU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Human GFAP (Glial Fibrillary Acidic Protein). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.056 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

GFAP is an intermediate filament (IF) protein that is found in glial cells such as astrocytes. It is closely related to its non-epithelial family members, vimentin, desmin, and peripherin, which are all involved in the structure and function of the cell’s cytoskeleton. GFAP helps to maintain astrocyte mechanical strength, as well as the shape of cells.GFAP is expressed in the central nervous system in astrocyte cells. It is involved in many cellular functioning processes, such as cell structure and movement, cell communication, and the functioning of the blood brain barrier. The predicted amino acid sequence indicated that GFAP shares structural similarities particularly in the central rod domain and to a lesser degree in the carboxyl-terminal domain with other IF proteins found in nonepithelial cell types.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.