Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Human GAL3 ELISA Kit

Human GAL3 ELISA Kit

96 wells, 1 kit
Catalog # MSPP-SEA303HU
Marketsource
Supplier:   Cloud-Clone
Supplier #:  SEA303HU
Human GAL3 ELISA Kit
Catalog # MSPP-SEA303HU
Supplier:  Cloud-Clone
Marketsource
CAS Number:  
Human GAL3 ELISA Kit
Human GAL3 ELISA Kit

96 wells, 1 kit

CAS Number:  
Price
$630.00
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

This assay has high sensitivity and excellent specificity for detecting Human GAL3 (Galectin 3). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.054 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

Galectin-3 contains a carboxyl-terminal lectin domain and an amino-terminal non-lectin part consisting primarily of short tandem repeats. It is widely distributed in tissues and found in epithelial cells, fibroblasts, dendritic cells, and inflammatory cells. In many cell types studied, galectin-3 is present diffusely in the cytoplasm, but is also localized to the nucleus and subcellular structures, such as mitochondria, phagosomes and exosomes, under specific conditions. It is secreted by various cell types, including monocytes, macrophages, and epithelial cells, and the extracellular protein can bind to a large number of different glycoconjugates on the cell surfaces and extracellular matrices.

 : For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Documents

Specifications

Specifications

  • Size:
    1 kit
  • No. of tests:
    96 wells
  • Primary antibody reactivity:
    Human
  • Detection Range:
    0.156 - 10 ng/ml
  • Sample Volume:
    100 µl
  • Host:
    Rabbit
  • Time to Results:
    3 h
  • Detection Method:
    Colorimetric
  • Cat. No.:
    MSPP-SEA303HU
  • Regulatory Status:
    RUO
  • Assay duration:
    Multiple steps
  • Sensitivity:
    0.054 ng/ml
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Assay Type (ELISA with LOV):
    Sandwich
  • Target protein:
    GAL3
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Shelf Life:
    12 Months
  • Cross Reactivity:
    No significant cross-reactivity or interference between Galectin 3 (GAL3) and analogues was observed
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.