Order Entry
Puerto Rico
ContactUsLinkComponent
Human FGL1 ELISA Kit
Human FGL1 ELISA Kit
Catalog # MSPP-SED022HU
CAS Number:  
undefined
Human FGL1 ELISA Kit
Catalog # MSPP-SED022HU
Supplier Number:  SED022HU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Human
  • Target protein:
    FGL1
  • Size:
    1 kit
  • Sample Type:
    Plasma
  • Cross Reactivity:
    No significant cross-reactivity or interference between Fibrinogen Like Protein 1 (FGL1) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    12.5 - 800 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    4.4 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SED022HU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Human FGL1 (Fibrinogen Like Protein 1). The assay range is from 12.5 to 800 ng/ml (Sandwich kit) with a sensitivity of 4.4 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

Fibrinogen-like 1 is a member of the fibrinogen family. This protein is homologous to the carboxy terminus of the fibrinogen beta- and gamma- subunits which contains the four conserved cysteines of fibrinogens and fibrinogen related proteins. The deduced 312 amino acid FGL1 protein possesses a hydrophobic leader peptide. FGL1 shares significant sequence homology with FGB and FGG and with other fibrinogen-related proteins. The FGL1 sequence includes 4 cysteine residues that are widely conserved in all fibrinogens and fibrinogen-related proteins. However, this protein lacks the platelet-binding site, cross-linking region and a thrombin-sensitive site which are necessary for fibrin clot formation. This protein may play a role in the development of hepatocellular carcinomas.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.