Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Human FABP1 ELISA Kit

Human FABP1 ELISA Kit

96 wells, 1 kit
Catalog # MSPP-SEB566HU
Marketsource
Supplier:   Cloud-Clone
Supplier #:  SEB566HU
Human FABP1 ELISA Kit
Catalog # MSPP-SEB566HU
Supplier:  Cloud-Clone
Marketsource
CAS Number:  
Human FABP1 ELISA Kit
Human FABP1 ELISA Kit

96 wells, 1 kit

CAS Number:  
Price
$665.00
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

This assay has high sensitivity and excellent specificity for detecting Human FABP1 (Fatty Acid Binding Protein 1). The assay range is from 1.56 to 100 ng/ml (Sandwich kit) with a sensitivity of 0.59 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

FABP1 encodes the fatty acid binding protein found in liver. Fatty acid-binding protein is an abundant constituent of the cytoplasm that regulates lipid transport and metabolism. It binds free fatty acids, their CoA derivatives, bilirubin, organic anions, and other small molecules. Liver fatty acid-binding protein is required for cholesterol synthesis and metabolism. Chen et al. (1986) used cDNA of liver FABP in Southern analysis of human/mouse cell hybrid DNA and assigned the FABPL locus to 2p. Sparkes et al. (1987) confirmed the assignment to chromosome 2 and regionalized the gene to 2p11 by in situ hybridization. Sweetser et al. (1987) found that the mouse homolog of this gene is located on chromosome 6, within 3 cm of the lymphocyte antigen-2 (Ly-2) locus.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Documents

Specifications

Specifications

  • Size:
    1 kit
  • No. of tests:
    96 wells
  • Primary antibody reactivity:
    Human
  • Detection Range:
    1.56 - 100 ng/ml
  • Sample Volume:
    100 µl
  • Host:
    Rabbit
  • Time to Results:
    3 h
  • Detection Method:
    Colorimetric
  • Cat. No.:
    MSPP-SEB566HU
  • Regulatory Status:
    RUO
  • Assay duration:
    Multiple steps
  • Sensitivity:
    0.59 ng/ml
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Assay Type (ELISA with LOV):
    Sandwich
  • Target protein:
    FABP1
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Shelf Life:
    12 Months
  • Cross Reactivity:
    No significant cross-reactivity or interference between Fatty Acid Binding Protein 1 (FABP1) and analogues was observed
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.