Order Entry
Puerto Rico
ContactUsLinkComponent
Human ES ELISA Kit
Human ES ELISA Kit
Catalog # MSPP-SEA542HU
CAS Number:  
undefined
Human ES ELISA Kit
Catalog # MSPP-SEA542HU
Supplier Number:  SEA542HU
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Human
  • Target protein:
    ES
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Endostatin (ES) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    0.625 - 40 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    0.255 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEA542HU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Human ES (Endostatin). The assay range is from 0.625 to 40 ng/ml (Sandwich kit) with a sensitivity of 0.255 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

Endostatin is a broad spectrum angiogenesis inhibitor and may interfere with the pro-angiogenic action of growth factors Endostatin is a natural antiangiogenic protein that has been shown to inhibit the growth of blood vessels, thereby starving cancerous tumors. In preclinical studies, the repeated administration of Endostatin consistently shrank primary tumors and, unlike traditional chemotherapy, showed no drug resistance over time. Endostatin is currently being studied as part of cancer research. In a Phase I clinical trial of Endostatin of the 19 patients treated, 12 were switched out of the trial by their physicians due to continued progression of their disease. In a Phase II clinical trial of Endostatin forty two patients with pancreatic endocrine tumors or carcinoid tumors were treated. A phase III clinical trial was carried out on 493 histology or cytology confirmed stage IIIB and IV NSCLC patients with a life expectancy >3 months.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.