Order Entry
Puerto Rico
ContactUsLinkComponent
Human COMP ELISA Kit
Human COMP ELISA Kit
Catalog # MSPP-SEB197HU
CAS Number:  
undefined
Human COMP ELISA Kit
Catalog # MSPP-SEB197HU
Supplier Number:  SEB197HU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Human
  • Target protein:
    COMP
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Cartilage Oligomeric Matrix Protein (COMP) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    3.12 - 200 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    1.21 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEB197HU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Human COMP (Cartilage Oligomeric Matrix Protein). The assay range is from 3.12 to 200 ng/ml (Sandwich kit) with a sensitivity of 1.21 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

Cartilage oligomeric matrix protein (COMP) is a member of the thrombospondin family of extracellular matrix proteins. The protein consists of five 87-kDa subunits held together by interchain disulfide bonds, forming a 435-kDa pentameric protein. COMP is expressed in all types of cartilage, vitreous of the eye, tendon, and vascular smooth muscle cells. Immunohistological staining of articular cartilage has revealed a developmentally regulated localization of COMP to the chondrocyte territorial and interterritorial matrix. COMP binds in a zinc-dependent manner to collagen type I and type II and also to collagen type IX. The thrombospondin family currently includes five members: TSP-1, TSP-2, TSP-3, TSP-4, and COMP (also referred to as TSP-5).

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.