Human COL4a1 ELISA Kit
This assay has high sensitivity and excellent specificity for detecting Human COL4a1 (Collagen Type IV Alpha 1). The assay range is from 4.7 to 300 ng/ml (Sandwich kit) with a sensitivity of 2.1 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
- High sensitivity and specificity
- Perfect reproducibility and consistency across batches
- Quality control with three-level inspections
- Wide range of targets/species available
- Intra-assay: CV<10%; Inter-assay: CV<12%
Collagen, type IV, alpha 1 is a major type IV alpha collagen chain of basement membranes. Like the other members of the type IV collagen gene family, this gene is organized in a head-to-head conformation with another type IV collagen gene so that each gene pair shares a common promoter. Types I, II, and III collagen, the so-called interstitial collagens, are in many ways distinct from basement membrane collagen. Type IV collagen does not form ordered fibrillar structures; rather, a meshwork is formed by 4 molecules held together at the ends. Both disulfide and typical lysyl-derived collagen crosslinks are involved. The collagen IV molecule is a heterotrimer of 2 alpha-1 chains and 1 alpha-2 chain. The alpha-2 chain is encoded at a distinct locus (COL4A2).
: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.
- Size:1 kit
- No. of tests:96 wells
- Primary antibody reactivity:Human
- Detection Range:4.7 - 300 ng/ml
- Sample Volume:100 µl
- Host:Rabbit
- Time to Results:3 h
- Detection Method:Colorimetric
- Cat. No.:MSPP-SEA149HU
- Regulatory Status:RUO
- Assay duration:Multiple steps
- Sensitivity:2.1 ng/ml
- Conjugate ELISA:Biotin
- Format:Pre-coated
- Assay Type (ELISA with LOV):Sandwich
- Target protein:COL4a1
- Sample Type:Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
- Shelf Life:12 Months
- Cross Reactivity:No significant cross-reactivity or interference between Collagen Type IV Alpha 1 (COL4a1) and analogues was observed
- Storage Temperature:4 °C for one month (frequent use), −20 °C for one year



