Order Entry
Puerto Rico
ContactUsLinkComponent
Human CLU ELISA Kit
Human CLU ELISA Kit
Catalog # MSPP-SEB180HU
CAS Number:  
undefined
Human CLU ELISA Kit
Catalog # MSPP-SEB180HU
Supplier Number:  SEB180HU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Human
  • Target protein:
    CLU
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma, urine, saliva, tissue homogenates, cerebrospinal fluid, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Clusterin (CLU) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    12.5 - 800 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    5.1 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEB180HU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Human CLU (Clusterin). The assay range is from 12.5 to 800 ng/ml (Sandwich kit) with a sensitivity of 5.1 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

Clusterin (apolipoprotein J) is a 75 - 80 kDa disulfide-linked heterodimeric protein associated with the clearance of cellular debris and apoptosis. This protein has several synonyms: dimeric acidic glycoprotein (DAG protein), testosterone repressed prostate message-2 (TRPM-2), sulfated glycoprotein-2 (SGP-2) and complement lysis inhibitor (CLI).This protein was first identified in 1988 in seminal fluid.In humans the gene is encoded on chromosome 8 (8p21) and is highly conserved between species (70-80% homology).It is expressed in most mammalian tissues and can be found in blood plasma, milk, urine, cerebrospinal fluid and semen. A number of proteins have been found to affect its expression including Egr-1, members from the AP-1 complex, HSF1/2, Cdx1/2 and B-Myb.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.