Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human ADAM10 ELISA Kit
Catalog # MSPP-SEA766HU
CAS Number:  
undefined
Human ADAM10 ELISA Kit
Catalog # MSPP-SEA766HU
Supplier Number:  SEA766HU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Human
  • Target protein:
    ADAM10
  • Size:
    1 kit
  • Sample Type:
    Tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between A Disintegrin And Metalloprotease 10 (ADAM10) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    78 - 5000 pg/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    30 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEA766HU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Human ADAM10 (A Disintegrin And Metalloprotease 10). The assay range is from 78 to 5000 pg/ml (Sandwich kit) with a sensitivity of 30 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

A Disintegrin And Metalloprotease 10 is a ADAM family member that cleaves many proteins including TNF-alpha and E-cadherin. Members of the ADAM family are cell surface proteins with a unique structure possessing both potential adhesion and protease domains. ADAM10 is a sheddase, and has a broad specificity for peptide hydrolysis reactions. New data on the ADAM10 shedding of ephrin/eph complex associated with Eph on one cell surface has determined that ADAM10 cleaves ephrin, within the ephrin/eph complex, formed between two cell surfaces. When ephrin is freed from the opposing cell, the entire ephrin/eph complex is endocytosed. This shedding in trans had not been previously shown, but may well be involved in other shedding events.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.