Order Entry
Puerto Rico
ContactUsLinkComponent
Human ADA ELISA Kit
Human ADA ELISA Kit
Catalog # MSPP-SEB390HU
CAS Number:  
undefined
Human ADA ELISA Kit
Catalog # MSPP-SEB390HU
Supplier Number:  SEB390HU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Human
  • Target protein:
    ADA
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Adenosine Deaminase (ADA) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    0.156 - 10 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    0.061 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEB390HU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Human ADA (Adenosine Deaminase). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.061 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

Adenosine deaminase is an enzyme (EC 3.5.4.4) involved in purine metabolism. It is needed for the breakdown of adenosine from food and for the turnover of nucleic acids in tissues. Plays an important role in purine metabolism and in adenosine homeostasis. Modulates signaling by extracellular adenosine, and so contributes indirectly to cellular signaling events. Acts as a positive regulator of T-cell coactivation, by binding DPP4. Its interaction with DPP4 regulates lymphocyte-epithelial cell adhesion. ADA exists in both small form (as a monomer) and large form (as a dimer-complex). In the monomer form, the enzyme is a polypeptide chain, folded into eight strands of parallel α/β barrels, which surround a central deep pocket that is the active site. In addition to the eight central β-barrels and eight peripheral α-helices, ADA also contains five additional helices: residues 19-76 fold into three helices, located between β1 and α1 folds; and two antiparallel carboxy-terminal helices are located across the amino-terminal of the β-barrel. Its primary function in humans is the development and maintenance of the immune system. However, the full physiological role of ADA is not yet completely understood.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.