Order Entry
Puerto Rico
ContactUsLinkComponent
Guinea pig IL4 ELISA Kit
Guinea pig IL4 ELISA Kit
Catalog # MSPP-SEA077GU
CAS Number:  
undefined
Guinea pig IL4 ELISA Kit
Catalog # MSPP-SEA077GU
Supplier Number:  SEA077GU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Guinea pig
  • Target protein:
    IL4
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Interleukin 4 (IL4) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    3.12 - 200 pg/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    1.14 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEA077GU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Guinea pig IL4 (Interleukin 4). The assay range is from 3.12 to 200 pg/ml (Sandwich kit) with a sensitivity of 1.14 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

Interleukin 4 is a pleiotropic cytokine produced primarily by activated T lymphocytes,mast cells and basophils. IL-4 has multiple immune response-modulating functions on a variety of cell types. It is an important modulator of the differentiation of precursor T helper cells to the Th2 subset that mediates humoral immunity and modulates antibody production. In addition, IL-4 has also been shown to have anti-tumor activity both in vivoand in vitro.The biological effects of IL-4 are mediated by specific cell surface receptor complexes. One type of functional IL-4 receptor complex consists of the IL-4-binding subunit and a second chain, designated the common c chain because it has also been identified as a component of the receptor complexes for IL-4, IL-7, IL-9 and IL-15.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.