Order Entry
Puerto Rico
ContactUsLinkComponent
General Species VB6 ELISA Kit
General Species VB6 ELISA Kit
Catalog # MSPP-CEA916GE
Supplier:  Cloud-Clone
CAS Number:  
undefined
General Species VB6 ELISA Kit
Catalog # MSPP-CEA916GE
Supplier:  Cloud-Clone
Supplier Number:  CEA916GE
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Competitive
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Multiple
  • Target protein:
    VB6
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Vitamin B6 (VB6) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    2 h
  • Shelf Life:
    12 Months
  • Detection Range:
    3.70 - 300 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    50 µl
  • Sensitivity:
    1.42 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-CEA916GE
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting General species VB6 (Vitamin B6). The assay range is from 3.70 to 300 ng/ml (Competitive kit) with a sensitivity of 1.42 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

Vitamin B6 is a water-soluble vitamin. Pyridoxine is one of the compounds that can be called vitamin B6, along with pyridoxal and pyridoxamine. Its hydrochloride salt pyridoxine hydrochloride is often used.  Vitamin B6 assists in the balancing of sodium and potassium as well as promoting red blood cell production. It is linked to cardiovascular health by decreasing the formation of homocysteine. Pyridoxine may help balance hormonal changes in women and aid the immune system. It is required for the production of the monoamine neurotransmitters serotonin, dopamine, norepinephrine and epinephrine, as it is the precursor to pyridoxal phosphate: cofactor for the enzyme aromatic amino acid decarboxylase. This enzyme is responsible for converting the precursors 5-hydroxytryptophan (5-HTP) into serotonin and melatonin, and levodopa (L-DOPA) into dopamine, noradrenaline and adrenaline.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.