Order Entry
Puerto Rico
ContactUsLinkComponent
General Species TCA ELISA Kit
General Species TCA ELISA Kit
Catalog # MSPP-CEO267GE
CAS Number:  
undefined
General Species TCA ELISA Kit
Catalog # MSPP-CEO267GE
Supplier Number:  CEO267GE
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Competitive
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Multiple
  • Target protein:
    TCA
  • Size:
    1 kit
  • Sample Type:
    Biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Taurocholic Acid (TCA) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    2 h
  • Shelf Life:
    12 Months
  • Detection Range:
    3.70 - 300 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    50 µl
  • Sensitivity:
    1.33 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-CEO267GE
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting General species TCA (Taurocholic Acid). The assay range is from 3.70 to 300 ng/ml (Competitive kit) with a sensitivity of 1.33 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

Taurocholic acid is a deliquescent yellowish crystalline bile acid involved in the emulsification of fats. It occurs as a sodium salt in the bile of mammals. It is a conjugate of cholic acid with taurine. In medical use, it is administered as a cholagogue and choleretic. Hydrolysis of taurocholic acid yields taurine. For commercial use, taurocholic acid is manufactured from cattle bile, a byproduct of the meat-processing industry. This acid is also one of the many molecules in the body that has cholesterol as its precursor. Taurocholic acid absorption was monitored as changes in short-circuit current (SCC) in intact and stripped ileal sheets from normal (Swiss) and transgenic CF (Cftr(tm2Cam)) mice with the DeltaF508 mutation. Taurocholic acid uptake was measured directly in everted ileal sacs and in brush-border membrane vesicles (BBMVs) using radiolabelled bile acid.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.