Order Entry
United States
ContactUsLinkComponent
 
General Species PAF ELISA Kit
Catalog # MSPP-CEA526GE
CAS Number:  
undefined
General Species PAF ELISA Kit
Catalog # MSPP-CEA526GE
Supplier Number:  CEA526GE
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Competitive
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Multiple
  • Target protein:
    PAF
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Platelet Activating Factor (PAF) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    2 h
  • Shelf Life:
    12 Months
  • Detection Range:
    37.0 - 3000 pg/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    50 µl
  • Sensitivity:
    13.3 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-CEA526GE
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting General species PAF (Platelet Activating Factor). The assay range is from 37.0 to 3000 pg/ml (Competitive kit) with a sensitivity of 13.3 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

Platelet-activating factor is a potent phospholipid activator and mediator of many leukocyte functions, including platelet aggregation, inflammation, and anaphylaxis.It is produced in response to specific stimuli by a variety of cell types, including neutrophils, basophils, platelets, and endothelial cells.Several molecular species of platelet-activating factor have been identified which vary in the length of the O-alkyl side chain.It is an important mediator of bronchoconstriction.It causes platelets to aggregate and blood vessels to dilate. Thus it is important to the process of hemostasis. At a concentration of 10^-12 M, PAF causes life threatening inflammation of the airways to induce asthma like symptoms.Toxins such as fragments of destroyed bacteria induce the synthesis of PAF, which causes a drop in blood pressure and reduced volume of blood pumped by the heart, which leads to shock and maybe death.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.