Order Entry
Puerto Rico
ContactUsLinkComponent
General Species HVA ELISA Kit
General Species HVA ELISA Kit
Catalog # MSPP-CED717GE
CAS Number:  
undefined
General Species HVA ELISA Kit
Catalog # MSPP-CED717GE
Supplier Number:  CED717GE
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Competitive
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Multiple
  • Target protein:
    HVA
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Homovanillic Acid (HVA) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    2 h
  • Shelf Life:
    12 Months
  • Detection Range:
    2.47 - 200 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    50 µl
  • Sensitivity:
    0.95 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-CED717GE
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting General species HVA (Homovanillic Acid). The assay range is from 2.47 to 200 ng/ml (Competitive kit) with a sensitivity of 0.95 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

Homovanillic acid (HOC₆H₃(OCH₃)CH₂COOH; synonyms: 3-Methoxy-4-hydroxyphenyl acetic acid; HVA; 4-Hydroxy-3-methoxy-benzeneacetic acid; 4-Hydroxy-3-methoxyphenylacetic acid) is a major catecholamine metabolite. It is used as a reagent to detect oxidative enzymes, and is associated with dopamine levels in the brain. In psychiatry and neuroscience, brain and cerebrospinal fluid levels of HVA are measured as a marker of metabolic stress caused by 2-deoxy-D-glucose. HVA presence supports a diagnosis of neuroblastoma and malignant pheochromocytoma. An acid that is produced by the normal metabolism of dopamine and that may occur at an elevated level in urine in association with tumors of the adrenal gland. Its normal accumulation in a 24-hour collection urine sample is 15 mg.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.