- Pack type:Vial
- Conjugation:Unconjugated
- Protein Function:Cytokines and Growth Factors
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:50 µg
- Tag sequence:MGHHHHHHSGSEF
- Storage Conditions:–20 °C
- Endotoxin Content:<1.0 EU per 1 μg (determined by the LAL method)
- Gene ID:3326
- Reconstitution Instructions:Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:Heat Shock Protein 90kDa Alpha B1
- UniProtKB:P08238
- Protein/Peptide Name:HSP90aB1
- Purity:97 - 100%
- Molecular Weight:81 kDa
- Sequence:Met1~Asp724
- Endotoxin Level:Low
- Concentration:0.2 mg/ml
- Formulation:Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose.
- Nuclease-free:N
- Shipping Temperature:4 °C
- Tested Applications:Positive control, Immunogen, SDS-PAGE, Western blot.
- Cat. No.:MSPP-RPD522HU1
This is a HSP90aB1 recombinant protein (prokaryotic), Human is sequencing from Met1~Asp724 with 97 - 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
- High quality, purity, reproducibility and effectiveness
- Offers customized buffers and tag options
- 100% quality and service satisfaction guarantee
HSP90 proteins are highly conserved molecular chaperones that have key roles in signal transduction, protein folding, protein degradation, and morphologic evolution. There are 2 major cytosolic HSP90 proteins, HSP90AA1, an inducible form, and HSP90AB1, a constitutive form. Other HSP90 proteins are found in endoplasmic reticulum (HSP90B1) and mitochondria (TRAP1).Like other HSP90 proteins, the 724-amino acid HSP90AB1 protein has a highly conserved N-terminal domain, a charged domain, a middle domain involved in ATPase activity, a second charged domain, and a C-terminal domain. It also has a 4-helical cytokine motif, a gln-rich region, and a C-terminal MEEVD motif characteristic of cytosolic HSP90 proteins.