- Pack type:Vial
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:50 µg
- Tag sequence:MGHHHHHHSGSEF
- Storage Conditions:–20 °C
- Endotoxin Content:<1.0 EU per 1 μg (determined by the LAL method)
- Gene ID:10551
- Reconstitution Instructions:Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:Anterior Gradient 2
- UniProtKB:O95994
- Protein/Peptide Name:AGR2
- Purity:90 - 100%
- Molecular Weight:22 kDa
- Sequence:Arg21~Leu175
- Endotoxin Level:Low
- Concentration:0.2 mg/ml
- Formulation:Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose.
- Nuclease-free:N
- Shipping Temperature:4 °C
- Tested Applications:Positive control, Immunogen, SDS-PAGE, Western blot.
- Cat. No.:MSPP-RPC285HU1
This is a AGR2 recombinant protein (prokaryotic), Human is sequencing from Arg21~Leu175 with 90 - 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
- High quality, purity, reproducibility and effectiveness
- Offers customized buffers and tag options
- 100% quality and service satisfaction guarantee
Anterior gradient homolog 2 (AGR2) is a human protein originally discovered in Xenopus laevis, encoded for by the gene of the same name. In Xenopus AGR2 plays a role in cement gland differentiation, however in human cancer cell lines high levels of AGR2 correlate with downregulation of the p53 response and AGR2 levels are elevated in the preneoplastic tissue Barrett's oesophagus. The deduced 175-amino acid soluble AGR2 protein, which is 91% identical to the mouse protein and 47% identical to the frog protein, contains a signal peptide. Northern blot analysis revealed strongest expression of 0.9- and 1.6-kb AGR2 transcripts in lung and in all ER-positive breast carcinoma lines tested; weaker expression was detected in pancreas.