Order Entry
Puerto Rico
ContactUsLinkComponent
Human Synthetic OxLDL (from Synthetic)
Human Synthetic OxLDL (from Synthetic)
Catalog # MSPP-SPA527HU1
CAS Number:  
Human Synthetic OxLDL (from Synthetic)
Catalog # MSPP-SPA527HU1
Supplier Number:  SPA527HU01
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/Peptide Type:
    Synthetic
  • Source:
    Synthetic
  • Species:
    Human
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <1.0 EU per 1 μg (determined by the LAL method)
  • Gene ID:
    338
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Oxidized Low Density Lipoprotein
  • UniProtKB:
    P04114
  • Protein/Peptide Name:
    OxLDL
  • Purity:
    95 - 100%
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose.
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Immunogen, Blocking peptide.
  • Cat. No.:
    MSPP-SPA527HU1

Specifications

About this item

This is a OxLDL synthetic peptides, Human is sequencing from with 95 - 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

Hypercholesterolemia is closely associated with increased risk of atherosclerosis. The cholesterol accumulated in the atherosclerotic plaque is derived primarily from low-density lipoprotein (LDL). Oxidation of LDL is now accepted as a critical event in the atherogenic process. Free radicals like superoxide and nitric oxide (-O2 , NO) generated in biological reactions in the body contribute to the oxidation of LDL. The NO radical has been shown to oxidize apolipoprotein B a constituant of LDL. Similarly, lipoxygenases and oxidants like peroxynitrite can oxidize the lipid moieties in LDL. It has been demonstrated that oxLDL and not native LDL is taken up by scavenger receptors on monocytes, smooth muscle cells and macrophages in the blood vessels. The oxidation of LDL increases the affinity of oxLDL to acetyl receptors of macrophages. As this pathway of oxLDL uptake is unregulated, it results in the formation of lipid-laden macrophages (foam cells).

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.