Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant SRSF2 (Prokaryotic) (from E. coli)
Human Recombinant SRSF2 (Prokaryotic) (from E. coli)
Catalog # MSPP-RPF102HU1
Supplier:  Cloud-Clone
CAS Number:  
undefined
Human Recombinant SRSF2 (Prokaryotic) (from E. coli)
Catalog # MSPP-RPF102HU1
Supplier:  Cloud-Clone
Supplier Number:  RPF102HU01
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    50 µg
  • Tag sequence:
    MSPILGYWKIKGLVQPTRLLLEYLEEKYEEHLYERDEGDKWRNKKFELGLEFPNLPYYIDGDVKLTQSMAIIRYIADKHNMLGGCPKERAEISMLEGAVLDIRYGVSRIAYSKDFETLKVDFLSKLPEMLKMFEDRLCHKTYLNGDHVTHPDFMLYDALDVVLYMDPMCLDAFPKLVCFKKRIEAIPQIDKYLKSSKYIAWPLQGWQATFGGGDHPPKSDGSTSGSGHHHHHHSAGLVPRGSTAIGMKETAAAKFERQHMDSPDLGTGGGSGIEGRGSMGYRGSEF
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <1.0 EU per 1 μg (determined by the LAL method)
  • Gene ID:
    6427
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Serine/Arginine Rich Splicing Factor 2
  • UniProtKB:
    Q01130
  • Protein/Peptide Name:
    SRSF2
  • Purity:
    90 - 100%
  • Molecular Weight:
    54 kDa
  • Sequence:
    Thr14~Ser221
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose.
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Positive control, Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-RPF102HU1

Specifications

About this item

This is a SRSF2 recombinant protein (prokaryotic), Human is sequencing from Thr14~Ser221 with 90 - 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

SFRS2 contains a ribonucleoprotein (RNP)-type RNA-binding motif and a carboxyl-terminal serine/arginine-rich (SR) domain.in every member of the human SR family of splicing regulators, highly or ultraconserved elements are alternatively spliced, either as alternative 'poison cassette exons' containing early in-frame stop codons, or as alternative introns in the 3-prime untranslated region. These alternative splicing events target the resulting mRNAs for degradation by means of an RNA surveillance pathway called nonsense-mediated mRNA decay. Mouse orthologs of the human SR proteins exhibit the same unproductive splicing patterns. Three SR proteins, SRp20, SC35, and 9G8 (SFRS7), had been previously shown to direct splicing of their own transcripts, and SC35 autoregulates its expression by coupling alternative splicing with decay.

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.