Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Recombinant GAL3 (Prokaryotic) (from E. coli)
Mouse Recombinant GAL3 (Prokaryotic) (from E. coli)
Catalog # MSPP-RPA303MU1
Supplier:  Cloud-Clone
CAS Number:  
undefined
Mouse Recombinant GAL3 (Prokaryotic) (from E. coli)
Catalog # MSPP-RPA303MU1
Supplier:  Cloud-Clone
Supplier Number:  RPA303MU01
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <1.0 EU per 1 μg (determined by the LAL method)
  • Gene ID:
    16854
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Galectin 3
  • UniProtKB:
    Q8C253
  • Protein/Peptide Name:
    GAL3
  • Purity:
    97 - 100%
  • Molecular Weight:
    31 kDa
  • Sequence:
    Ala2~Ile264
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose.
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Positive control, Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-RPA303MU1

Specifications

About this item

This is a GAL3 recombinant protein (prokaryotic), Mouse is sequencing from Ala2~Ile264 with 97 - 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

Galectin-3 contains a carboxyl-terminal lectin domain and an amino-terminal non-lectin part consisting primarily of short tandem repeats. It is widely distributed in tissues and found in epithelial cells, fibroblasts, dendritic cells, and inflammatory cells. In many cell types studied, galectin-3 is present diffusely in the cytoplasm, but is also localized to the nucleus and subcellular structures, such as mitochondria, phagosomes and exosomes, under specific conditions. It is secreted by various cell types, including monocytes, macrophages, and epithelial cells, and the extracellular protein can bind to a large number of different glycoconjugates on the cell surfaces and extracellular matrices.

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.