- Pack type:Vial
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:50 µg
- Tag sequence:MGHHHHHHSGSEF
- Storage Conditions:–20 °C
- Endotoxin Content:<1.0 EU per 1 μg (determined by the LAL method)
- Gene ID:5806
- Reconstitution Instructions:Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:Pentraxin 3, Long
- UniProtKB:P26022
- Protein/Peptide Name:PTX3
- Purity:95 - 100%
- Molecular Weight:26 kDa
- Sequence:Ile183~Ser381
- Endotoxin Level:Low
- Concentration:0.2 mg/ml
- Formulation:Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose.
- Nuclease-free:N
- Shipping Temperature:4 °C
- Tested Applications:Positive control, Immunogen, SDS-PAGE, Western blot.
- Cat. No.:MSPP-RPK411HU1
This is a PTX3 recombinant protein (prokaryotic), Human is sequencing from Ile183~Ser381 with 95 - 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
- High quality, purity, reproducibility and effectiveness
- Offers customized buffers and tag options
- 100% quality and service satisfaction guarantee
PTX3 bound selected pathogens, including Aspergillus fumigatus, Pseudomonas aeruginosa, and Salmonella typhimurium. PTX3 bound to either viable or heat-inactivated conidia, but not the hyphae, of A. fumigatus, and Ptx3 facilitated conidia interaction with mononuclear phagocytes. Conidia rapidly induced PTX3 production in human and mouse monocytes and dendritic cells, but not in neutrophils, fibroblasts, or endothelial or epithelial cells. Infection of mice and humans with A. fumigatus induced high levels of PTX3 in plasma and bronchoalveolar lavage fluid. Significant levels of PTX3 were detected in plasma of neutropenic patients with systemic A. fumigatus infection, but only low levels were found in control subjects.