Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant COL1a2 (Prokaryotic) (from E. coli)
Human Recombinant COL1a2 (Prokaryotic) (from E. coli)
Catalog # MSPP-RPA215HU1
CAS Number:  
undefined
Human Recombinant COL1a2 (Prokaryotic) (from E. coli)
Catalog # MSPP-RPA215HU1
Supplier Number:  RPA215HU01
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <1.0 EU per 1 μg (determined by the LAL method)
  • Gene ID:
    1278
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Collagen Type I Alpha 2
  • UniProtKB:
    P08123
  • Protein/Peptide Name:
    COL1a2
  • Purity:
    97 - 100%
  • Molecular Weight:
    33 kDa
  • Sequence:
    Val1103~Lys1366
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose.
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Positive control, Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-RPA215HU1

Specifications

About this item

This is a COL1a2 recombinant protein (prokaryotic), Human is sequencing from Val1103~Lys1366 with 97 - 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

Collagen alpha-2(I) chain is a protein, one of the chains for type I collagen, the fibrillar collagen found in most connective tissues. Mutations in this gene are associated with osteogenesis imperfecta, Ehlers-Danlos syndrome, idiopathic osteoporosis, and atypical Marfan syndrome. De Wet et al. (1987) isolated 60 kb of cloned DNA containing the entire COL1A2 gene and 22 kb of flanking sequences. Like the homologous avian gene, the 1,366 amino acid residues of the human prepropolypeptide chain are encoded by 52 exons. Analysis of the 5-prime and 3-prime untranslated regions conclusively established the nature of 5 polymorphic mRNA transcripts. The exons are equally distributed as follows: 6 in the N-propeptide domain, 42 in the alpha-chain region, and 4 in the C-propeptide domain.

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.