Order Entry
Puerto Rico
ContactUsLinkComponent
Bovine Recombinant IFNt (Prokaryotic) (from E. coli)
Bovine Recombinant IFNt (Prokaryotic) (from E. coli)
Catalog # MSPP-RPB862BO1
CAS Number:  
undefined
Bovine Recombinant IFNt (Prokaryotic) (from E. coli)
Catalog # MSPP-RPB862BO1
Supplier Number:  RPB862BO01
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Bovine
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <1.0 EU per 1 μg (determined by the LAL method)
  • Gene ID:
    317698
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Interferon Tau
  • UniProtKB:
    P15696
  • Protein/Peptide Name:
    IFNt
  • Purity:
    97 - 100%
  • Molecular Weight:
    23 kDa
  • Sequence:
    Cys24~Leu195
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose.
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Positive control, Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-RPB862BO1

Specifications

About this item

This is a IFNt recombinant protein (prokaryotic), Bovine is sequencing from Cys24~Leu195 with 97 - 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

Interferon tau is a Type I interferon made up of a single chain of amino-acids. In many species of ruminant, it acts as a signaling molecule during pregnancy. It is secreted by the trophoblast cells into the uterine lumen in days 13 to 21 of pregnancy. It decreases endometrial oxytocin receptors, which then cannot stimulate PGF-2-alpha synthesis, preventing luteolysis. It promotes uterine implantation by increasing protein synthesis in glands. Although its role in humans is not certain, it has been found to bind to the same receptors sensitive to interferon alpha, and thus may have anti-viral properties, as hinted by studies showing its ability to inhibit the reverse transcriptase enzyme found in retroviruses such as HIV. Its low toxicity compared to other interferons also supports its investigation as a potential therapeutic compound.

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.