Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant CK18 (Prokaryotic) (from E. coli)
Human Recombinant CK18 (Prokaryotic) (from E. coli)
Catalog # MSPP-RPB231HU1
CAS Number:  
undefined
Human Recombinant CK18 (Prokaryotic) (from E. coli)
Catalog # MSPP-RPB231HU1
Supplier Number:  RPB231HU01
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <1.0 EU per 1 μg (determined by the LAL method)
  • Gene ID:
    3875
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Cytokeratin 18
  • UniProtKB:
    P05783
  • Protein/Peptide Name:
    CK18
  • Purity:
    95 - 100%
  • Molecular Weight:
    29 kDa
  • Sequence:
    Asp238~Leu396
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose.
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Positive control, Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-RPB231HU1

Specifications

About this item

This is a CK18 recombinant protein (prokaryotic), Human is sequencing from Asp238~Leu396 with 95 - 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

Keratin 18 is a type I cytokeratin. It is, together with its filament partner keratin 8, perhaps the most commonly found products of the intermediate filament gene family. They are expressed in single layer epithelial tissues of the body. Mutations in this gene have been linked to cryptogenic cirrhosis. Two transcript variants encoding the same protein have been found for this gene. Human keratin 18 and the homologous mouse Endo B are type I IF protein subunits whose expression is restricted in adults to a variety of simple epithelial tissues. Endo B and its coexpressed type II keratin, Endo A (the mouse equivalent of human keratin 8), appear to be the first IF proteins expressed during murine development.

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.