Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human Recombinant WNT5A (Prokaryotic) (from E. coli)
Catalog # MSPP-RPP549HU1
CAS Number:  
undefined
Human Recombinant WNT5A (Prokaryotic) (from E. coli)
Catalog # MSPP-RPP549HU1
Supplier Number:  RPP549HU01
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <1.0 EU per 1 μg (determined by the LAL method)
  • Gene ID:
    7474
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Wingless Type MMTV Integration Site Family, Member 5A
  • UniProtKB:
    P41221
  • Protein/Peptide Name:
    WNT5A
  • Purity:
    90 - 100%
  • Molecular Weight:
    40 kDa
  • Sequence:
    Ile62~Lys380
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose.
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Positive control, Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-RPP549HU1

Specifications

About this item

This is a WNT5A recombinant protein (prokaryotic), Human is sequencing from Ile62~Lys380 with 90 - 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

WNT5A is a member of the WNT gene family. It encodes a protein which shows 98%, 98% and 87% amino acid identity to the mouse, rat and the xenopus Wnt5A protein, respectively. The experiments performed in Xenopus laevis embryos identified that human frizzled-5 (hFz5) is the receptor for the Wnt5A ligand and the Wnt5A/hFz5 signaling mediates axis induction.In the presence of a chemokine gradient, this Wnt-mediated receptor-actin-myosin polarity structure accumulates asymmetrically at the cell periphery, where it triggers membrane contractility and nuclear movement in the direction of membrane retraction. The process requires endosome trafficking, is associated with multivesicular bodies, and is regulated by Wnt5a through the small guanosine triphosphates Rab4 and RhoB.

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.