Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Recombinant PSA (Prokaryotic) (from E. coli)
Mouse Recombinant PSA (Prokaryotic) (from E. coli)
Catalog # MSPP-RPA151MU1
CAS Number:  
undefined
Mouse Recombinant PSA (Prokaryotic) (from E. coli)
Catalog # MSPP-RPA151MU1
Supplier Number:  RPA151MU01
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <1.0 EU per 1 μg (determined by the LAL method)
  • Gene ID:
    18050
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Prostate Specific Antigen
  • UniProtKB:
    P00756
  • Protein/Peptide Name:
    PSA
  • Purity:
    90 - 100%
  • Molecular Weight:
    27 kDa
  • Sequence:
    His39~Pro261
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose.
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Positive control, Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-RPA151MU1

Specifications

About this item

This is a PSA recombinant protein (prokaryotic), Mouse is sequencing from His39~Pro261 with 90 - 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

Human prostate-specific antigen (APS) is a kallikrein-like protease present in seminal plasma. It is a single-chain glycoprotein with a molecular mass of about 33 kDa that may function normally in the liquefaction of seminal coagulum. Radioimmunoassay of serum levels of this antigen (called PSA in the clinical setting) is useful in the diagnosis and monitoring of prostatic carcinoma. The full-length KLK3 protein contains 261 amino acids. It has a putative signal peptide, followed by a short activating peptide and the protease domain, which includes the catalytic triad of his65, asp120, and ser213. RT-PCR of 35 adult and fetal tissues detected highest expression in prostate. Expression was weak in colon, mammary gland, and parotid, and little to no expression was detected in other tissues examined.

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.