Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Recombinant Hsp60 (Prokaryotic) (from E. coli)
Mouse Recombinant Hsp60 (Prokaryotic) (from E. coli)
Catalog # MSPP-RPA822MU1
CAS Number:  
undefined
Mouse Recombinant Hsp60 (Prokaryotic) (from E. coli)
Catalog # MSPP-RPA822MU1
Supplier Number:  RPA822MU01
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <1.0 EU per 1 μg (determined by the LAL method)
  • Gene ID:
    15510
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Heat Shock Protein 60
  • UniProtKB:
    P63038
  • Protein/Peptide Name:
    Hsp60
  • Purity:
    90 - 100%
  • Molecular Weight:
    65 kDa
  • Sequence:
    Lys28~Phe573
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose.
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Positive control, Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-RPA822MU1

Specifications

About this item

This is a Hsp60 recombinant protein (prokaryotic), Mouse is sequencing from Lys28~pH e573 with 90 - 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

In control subjects no significant expression of IFN-γ or interleukin 4 was induced. These results suggest that periodontitis patients have human hsp60-reactive T cells with a type 1 cytokine profile in their peripheral blood T-cell pools.Heat shock protein 60 (HSP60) is homologous to HSP65 (a major antigenic mycobacterial protein) and E. coli GroEL. HSP60, also referred to as chaperonin-60, P1 and mitonin, has been classified as a member of a family of proteins termed chaperonins which act to recognize and stabilize polypeptide intermediates during folding, assembly and disassembly. HSP60 is an abundant protein which is constitutively expressed and is induced by environmental stress. HSP60 exists as a large oligomer composed of 14 to 60 kDa subunits arranged as two stacked rings.

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.