Order Entry
Puerto Rico
ContactUsLinkComponent
CheckMate/Flexi Vector Mammalian Two-Hybrid System, 1 each, Promega
CheckMate/Flexi Vector Mammalian Two-Hybrid System, 1 each, Promega
Catalog # PAC9360
CAS Number:  
undefined
CheckMate/Flexi Vector Mammalian Two-Hybrid System, 1 each, Promega
Catalog # PAC9360
Supplier Number:  C9360
CAS Number:  

Specifications

  • Description:
    CheckMate™/Flexi® Vector Mammalian Two-Hybrid System
  • Size:
    1 each
  • Cat. No.:
    PAC9360
  • Storage temperature:
    –30...–10 °C
  • Supplier no.:
    C9360

Specifications

About this item

The CheckMate/Flexi Vector Mammalian Two-Hybrid System provides a means to study suspected interactions between two proteins or domains and can also be used to generate stable cell lines for cell-based assays.

  • Study Mammalian Protein Interactions, Expression and Post-Translational Modifications
  • Confirm, validate and study suspected protein or domain interactions
  • Generate stable cell lines for cell-based assays
  • Enables mammalian protein expression and post-translational modification studies in situ

The CheckMate/Flexi Vector Mammalian Two-Hybrid System provides a means to confirm, validate and study suspected interactions between two proteins or domains and can also be used to generate stable cell lines for cell-based assays. Developed primarily for mammalian proteins of interest, the system can allow protein expression and post-translational modifications in an environment mimicking the native cell milieu. It is patterned on the yeast two-hybrid system with one protein of interest (X) fused to a DNA-binding domain and the other protein (Y) fused to a transcriptional activation domain. The system relies upon three plasmids that are co-transfected into mammalian cells, each plasmid having unique features. The pFN10A (ACT) Flexi Vector contains a herpes simplex virus VP16 transcriptional activation domain upstream of the cloning site, and the pFN11A (BIND) Flexi Vector contains the yeast GAL4DNA-binding domain upstream of the cloning site. The pFN11A (BIND) Flexi Vector also expresses the Renilla reniformis luciferase under the control of the SV40 promoter, allowing normalization for differences in transfection efficiency. The third vector, pGL4.31[luc2P/GAL4UAS/Hygro] Vector, contains five GAL4 binding sites upstream of a minimal TATA box, which is upstream of a firefly luciferase gene that acts as a reporter for interactions between proteins X and Y. This system differs from the original CheckMate Mammalian Two-Hybrid System in that the vectors are compatible with the Flexi Vector System, which allows directional cloning and rapid, efficient and high-fidelity transfer of protein coding regions between a variety of Flexi Vectors.

The vectors are compatible with the Flexi® Vector System, enabling convenient transfer of protein-coding regions for additional functional proteomics applications. All components should be stored at –20°C (–4°F).

Developed specifically for mammalian proteins, this system allows protein expression and post-translational modifications in an environment mimicking that of the native cell. It is patterned on the yeast two-hybrid system with one designated protein fused to a DNA-binding domain and the other protein fused to a transcriptional activation domain.

Ordering information: This system includes 5 µg of pFN10A (ACT) Flexi Vector, 5 µg of pFN11A (BIND) Flexi Vector, 20 µg of pGL4.31[luc2P/GAL4UAS/Hygro] Vector, 20 µg of pBIND Vector, 20 µg of pACT Vector, 20 µg of pBIND-Id Control Vector, 20 µg of pACT-MyoD Control Vector, and protocol.