Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant TIMP3 (prokaryotic)(from E. coli)
Human Recombinant TIMP3 (prokaryotic)(from E. coli)
Catalog # MSPP-RPA129HU1
CAS Number:  
undefined
Human Recombinant TIMP3 (prokaryotic)(from E. coli)
Catalog # MSPP-RPA129HU1
Supplier Number:  RPA129HU01
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <1.0 EU per 1 μg (determined by the LAL method)
  • Gene ID:
    7078
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Tissue Inhibitors Of Metalloproteinase 3
  • UniProtKB:
    P35625
  • Protein/Peptide Name:
    TIMP3
  • Purity:
    97 - 100%
  • Molecular Weight:
    26 kDa
  • Sequence:
    Cys26~Pro211
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4 containing 0.01% SKL, 5% trehalose
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Positive control, Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-RPA129HU1

Specifications

About this item

This is a TIMP3 recombinant protein (prokaryotic), Human is sequencing from Cys26~Pro211 with 97 to 100% purity. LyopH ilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

TIMP3,  The cDNAs contained a 591-bp open reading frame encoding 9 amino acid residues of the signal peptide and 188 residues of the mature TIMP3 polypeptide. Both the nucleotide sequence and the deduced translation product of the TIMP3 cDNA had a high degree of similarity to the TIMP1 and TIMP2 gene products, including 12 conserved cysteinyl residues at the same relative positions. The TIMP3 gene is expressed in many tissues, with highest expression in the placenta. Wilde et al. (1994) cloned and sequenced TIMP3 from phorbol ester-differentiated cells stimulated with bacterial lipopolysaccharide. The open reading frame encoded a 211-amino acid precursor, including a 23-residue secretion signal. The mature polypeptide had a calculated molecular weight of 21.6 kD.

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use In clinical diagnostic procedures. Please proper stored each component based on the instruction.