Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Recombinant cTnI (prokaryotic)(from E. coli)
Mouse Recombinant cTnI (prokaryotic)(from E. coli)
Catalog # MSPP-RPA478MU1
CAS Number:  
undefined
Mouse Recombinant cTnI (prokaryotic)(from E. coli)
Catalog # MSPP-RPA478MU1
Supplier Number:  RPA478MU01
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <1.0 EU per 1 μg (determined by the LAL method)
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Cardiac Troponin I
  • UniProtKB:
    P48787
  • Protein/Peptide Name:
    cTnI
  • Purity:
    97 - 100%
  • Molecular Weight:
    30 kDa
  • Sequence:
    Met1~Gly211
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4 containing 0.01% SKL, 5% trehalose
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Positive control, Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-RPA478MU1

Specifications

About this item

This is a cTnI recombinant protein (prokaryotic), Mouse is sequencing from Met1~Gly211 with 97 to 100% purity. LyopH ilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

Troponin I is part of a heteromeric complex playing an important role in the regulation of skeletal and cardiac muscle contraction. It consists of three subunits, troponin I (TnI), troponin T (TnT) and troponin C (TnC). Each subunit is responsible for part of troponin complex function. TnI inhibits ATPase activity of acto myosin and TnT and TnI are present in cardiac muscles in different forms than in skeletal muscles. Only one tissue specific isoform of TnI is described for cardiac muscle tissue (cTnI) and this is expressed only in myocardium.Equally reactive with free cardiac troponin I (cTnI) and cTnI forming complexes with other troponin components. (In the presence of 5 mM EDTA). Antibodies are not affected by heparin, phosphorylation, oxidation and troponin complex formation. No cross-reaction with skeletal muscle troponin I.

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use In clinical diagnostic procedures. Please proper stored each component based on the instruction.