Order Entry
Puerto Rico
ContactUsLinkComponent
Human Active IL13 (prokaryotic)(from E. coli)
Human Active IL13 (prokaryotic)(from E. coli)
Catalog # MSPP-APA060HU1
CAS Number:  
undefined
Human Active IL13 (prokaryotic)(from E. coli)
Catalog # MSPP-APA060HU1
Supplier Number:  APA060HU01
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <1.0 EU per 1 μg (determined by the LAL method)
  • Gene ID:
    3596
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Interleukin 13
  • UniProtKB:
    P35225
  • Protein/Peptide Name:
    IL13 active
  • Purity:
    95 - 100%
  • Molecular Weight:
    15 kDa
  • Sequence:
    Leu25~Asn146
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4 containing 0.01% SKL, 5% trehalose
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Cell culture, Activity Assays.
  • Cat. No.:
    MSPP-APA060HU1

Specifications

About this item

This is a IL13 active protein (prokaryotic), Human is sequencing from Leu25~Asn146 with 95 to 100% purity. LyopH ilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

Interleukin 13 (IL-13) is produced primarily by activated T lymphocytes. It shows approximately 30% amino acid (aa) sequence identity with IL-4 and shares many biological properties with IL-4. IL-13 has multiple effects on the differentiation and functions of monocytes/macrophages. It can suppress the cytotoxic functions of monocytes/macrophages and the production of proinflammatory cytokines by monocytes/macrophages. The biological effects of IL-13 are mediated by specific high-affinity cell surface receptor complexes. The functional IL-13 receptor complex has been shown to consist of the low-affinity IL-13 receptor α chain and the IL-4 receptor α subunit (IL-4 Rα). This IL-13 receptor complex may also serve as an alternate high-affinity IL-4 receptor complex in IL-4 responsive cells that lack the γ chain.

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use In clinical diagnostic procedures. Please proper stored each component based on the instruction.