- Pack type:Vial
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Mouse
- Size:50 µg
- Tag sequence:MGHHHHHHSGSEF
- Storage Conditions:–20 °C
- Endotoxin Content:<1.0 EU per 1 μg (determined by the LAL method)
- Reconstitution Instructions:Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:Pulmonary Surfactant Associated Protein A1
- UniProtKB:P35242
- Protein/Peptide Name:SFTPA1
- Purity:95 - 100%
- Molecular Weight:30 kDa
- Sequence:Asn21~Phe248
- Endotoxin Level:Low
- Concentration:0.2 mg/ml
- Formulation:Lyophilized from PBS, pH 7.4 containing 0.01% SKL, 5% trehalose
- Nuclease-free:N
- Shipping Temperature:4 °C
- Tested Applications:Positive control, Immunogen, SDS-PAGE, Western blot.
- Cat. No.:MSPP-RPA890MU1
This is a SFTPA1 recombinant protein (prokaryotic), Mouse is sequencing from Asn21~pH e248 with 95 to 100% purity. LyopH ilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.
- High quality, purity, reproducibility and effectiveness
- Offers customized buffers and tag options
- 100% quality and service satisfaction guarantee
SP-A (PRL) is a peptide hormone primarily associated with lactation. In breastfeeding, the infant suckling the teat stimulates the production of SP-A, which fills the breast with milk (lactogenesis) in preparation for the next feed. Oxytocin, a similar hormone, is also released, which triggers milk let-down. SP-A (PRL) is a polypeptide hormone secreted by anterior pituitary of both male and female. Release of SP-A is controlled by a complex neuroendocrine reflex initiated by a tactile stimulus and regulated by hypothalamic releasing and inhibition. Genetic variations in SFTPA1 are associated with respiratory distress syndrome in premature infants (RDS), also known as RDS in prematurity. RDS in the newborn is the main cause of mortality and morbidity in premature infants.