Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant PLA2R1 (prokaryotic)(from E. coli)
Human Recombinant PLA2R1 (prokaryotic)(from E. coli)
Catalog # MSPP-RPD851HU1
CAS Number:  
undefined
Human Recombinant PLA2R1 (prokaryotic)(from E. coli)
Catalog # MSPP-RPD851HU1
Supplier Number:  RPD851HU01
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <1.0 EU per 1 μg (determined by the LAL method)
  • Gene ID:
    22925
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Phospholipase A2 Receptor 1
  • UniProtKB:
    Q13018
  • Protein/Peptide Name:
    PLA2R1
  • Purity:
    97 - 100%
  • Molecular Weight:
    33 kDa
  • Sequence:
    Tyr385~Cys643
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4 containing 0.01% SKL, 5% trehalose
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Positive control, Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-RPD851HU1

Specifications

About this item

This is a PLA2R1 recombinant protein (prokaryotic), Human is sequencing from Tyr385~Cys643 with 97 to 100% purity. LyopH ilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

Sequence analysis revealed that the deduced 1,465-amino acid PLA2R1 transmembrane protein contains a 25-amino acid signal sequence, 15 potential N-glycosylation sites, an N-terminal cysteine-rich domain, a fibronectin-like type II domain, 8 carbohydrate recognition domains, and a short C-terminal cytoplasmic tail containing a consensus sequence (asn-pro-X-tyr) also found in bovine Pla2r1. A casein-kinase II phosphorylation site is also located in this C-terminal region. Ancian et al. (1995) identified a splice variant encoding a soluble form of PLA2R1. Quantitative PCR analysis suggested that the ratio for the transcription of membrane to soluble PLA2R1 is 1.6. Northern blot analysis detected strong expression of 6.5- and 5.4-kb PLA2R1 transcripts in kidney, with moderate expression in placenta, lung, and skeletal muscle.

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use In clinical diagnostic procedures. Please proper stored each component based on the instruction.