Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant APOB100 (prokaryotic)(from E. coli)
Human Recombinant APOB100 (prokaryotic)(from E. coli)
Catalog # MSPP-RPA603HU1
CAS Number:  
undefined
Human Recombinant APOB100 (prokaryotic)(from E. coli)
Catalog # MSPP-RPA603HU1
Supplier Number:  RPA603HU01
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <1.0 EU per 1 μg (determined by the LAL method)
  • Gene ID:
    338
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Apolipoprotein B100
  • UniProtKB:
    P04114
  • Protein/Peptide Name:
    APOB100
  • Purity:
    90 - 100%
  • Molecular Weight:
    26 kDa
  • Sequence:
    His3365~Glu3548
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4 containing 0.01% SKL, 5% trehalose
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Positive control, Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-RPA603HU1

Specifications

About this item

This is a APOB100 recombinant protein (prokaryotic), Human is sequencing from His3365~Glu3548 with 90 to 100% purity. LyopH ilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

Apolipoprotein B100 is a protein that plays a role in metabolism. It is a form of low density lipoprotein (LDL). Apo-B levels correlate with the risk of coronary disease. The protein occurs in the plasma in 2 main isoforms, APOB48 and APOB100. The first is synthesized exclusively by the small intestine, the second by the liver. Both isoforms are coded by APOB and by a single mRNA transcript larger than 16 kb. Apo-B100 is the major physiological ligand for the LDL receptor. Apo-B100 is a large monomeric protein, containing 4536 amino acids. Apo-B100 is synthesized in the liver and is required for the assembly of VLDL. It does not interchange between lipoprotein particles, as do the other apolipoproteins, and it is found in IDL and LDL after the removal of the Apo-A, E and C. Apo-B48 is present in chylomicrons and their remnants. It is essential for the intestinal absorption of dietary lipids.

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use In clinical diagnostic procedures. Please proper stored each component based on the instruction.