Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant CX43 (prokaryotic)(from E. coli)
Human Recombinant CX43 (prokaryotic)(from E. coli)
Catalog # MSPP-RPA277HU1
CAS Number:  
undefined
Human Recombinant CX43 (prokaryotic)(from E. coli)
Catalog # MSPP-RPA277HU1
Supplier Number:  RPA277HU01
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <1.0 EU per 1 μg (determined by the LAL method)
  • Gene ID:
    2697
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Connexin 43
  • UniProtKB:
    P17302
  • Protein/Peptide Name:
    CX43
  • Purity:
    90 - 100%
  • Molecular Weight:
    26 kDa
  • Sequence:
    Ser244~Ile382
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4 containing 0.01% SKL, 5% trehalose
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Positive control, Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-RPA277HU1

Specifications

About this item

This is a CX43 recombinant protein (prokaryotic), Human is sequencing from Ser244~Ile382 with 90 to 100% purity. LyopH ilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

Connexin 43 is a connexin gap junction found in several organs and cell types, but is most often found in the heart. Current data suggest CX43 is essential for the development of normal cardiac architecture and ventricular conduction. Two members of the connexin gene family, connexins 43 and 32, or GJB1, are abundantly expressed in the heart and liver, respectively. GAP43-like immunoreactivity in rat is mainly present in sympathetic and sensory nerve fibers as well as in perivascular nerve terminals. This peptide is axonally transported predominantly in sensory and adrenergic axonsSohl et al. (2003) stated that the mouse and human CX43 share 97% amino acid identity. Northern blot analysis detected variable expression of a 3.0-kb CX43 transcript in both mouse and human, with highest expression in heart.

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use In clinical diagnostic procedures. Please proper stored each component based on the instruction.