Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant VIM (prokaryotic)(from E. coli)
Human Recombinant VIM (prokaryotic)(from E. coli)
Catalog # MSPP-RPB040HU1
Supplier:  Cloud-Clone
CAS Number:  
undefined
Human Recombinant VIM (prokaryotic)(from E. coli)
Catalog # MSPP-RPB040HU1
Supplier:  Cloud-Clone
Supplier Number:  RPB040HU01
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <1.0 EU per 1 μg (determined by the LAL method)
  • Gene ID:
    7431
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Vimentin
  • UniProtKB:
    P08670
  • Protein/Peptide Name:
    VIM
  • Purity:
    97 - 100%
  • Molecular Weight:
    52 kDa
  • Sequence:
    Ser2~Glu466
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4 containing 0.01% SKL, 5% trehalose
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Positive control, Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-RPB040HU1

Specifications

About this item

This is a VIM recombinant protein (prokaryotic), Human is sequencing from Ser2~Glu466 with 97 to 100% purity. LyopH ilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

Vimentin is a type III intermediate filament (IF) protein that is expressed in mesenchymal cells. IF proteins are found in all metazoan cells as well as bacteria. IF, along with tubulin-based microtubules and actin-based microfilaments, comprise the cytoskeleton. All IF proteins are expressed in a highly developmentally-regulated fashion, vimentin is the major cytoskeletal component of mesenchymal cells. Because of this, vimentin is often used as a marker of mesenchymally-derived cells or cells undergoing an epithelial-to-mesenchymal transition (EMT) during both normal development and metastatic progression. A vimentin monomer, like all other intermediate filaments, has a central α-helical domain, capped on each end by non-helical amino (head) and carboxyl (tail) domains.

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use In clinical diagnostic procedures. Please proper stored each component based on the instruction.