Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Human Recombinant EIF4A1 (prokaryotic)(from <i>E. coli</i>)

Human Recombinant EIF4A1 (prokaryotic)(from E. coli)

Vial, 50 µg
Catalog # MSPP-RPF009HU1
Marketsource
Supplier:   Cloud-Clone
Supplier #:  RPF009HU01
Human Recombinant EIF4A1 (prokaryotic)(from <i>E. coli</i>)
Catalog # MSPP-RPF009HU1
Supplier:  Cloud-Clone
Marketsource
CAS Number:  
Human Recombinant EIF4A1 (prokaryotic)(from <i>E. coli</i>)
Human Recombinant EIF4A1 (prokaryotic)(from E. coli)

Vial, 50 µg

CAS Number:  
Price
$380.00
Log in for availability
Each(1/Itm/Bx)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

This is a EIF4A1 recombinant protein (prokaryotic), Human is sequencing from Met1~Thr271 with 95 to 100% purity. LyopH ilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

The eukaryotic initiation factor-4A family consists of 2 closely related genes, EIF4A1 and EIF4A2. These factors are required for the binding of mRNA to 40S ribosomal subunits. Nielsen et al. (1985) cloned eif4a1 cDNAs from mouse liver. They identified 2 distinct cDNAs differing in their untranslated regions. The sizes of these cDNAs correspond to 2 discrete mRNA bands of 2.0 and 1.6 kb seen on Northern blots of both mouse and human cells. Nielsen and Trachsel (1988) found that eif4a1 was expressed at similar levels in all mouse tissues examined, while eif4a2 had a much more varied pattern of expression. The EIF4A1 cDNA encodes a predicted 406-amino acid polypeptide with 92.7% amino acid similarity to the mouse protein. Kukimoto et al. (1997) characterized the promoter region of human EIF4A1.

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.

Caution: For research use only. Not for use In clinical diagnostic procedures. Please proper stored each component based on the instruction.

Documents

Specifications

Specifications

  • Pack type:
    Vial
  • Size:
    50 µg
  • Storage Conditions:
    –20 °C
  • Molecular Weight:
    35 kDa
  • Protease-free:
    N
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin Level:
    Low
  • UniProtKB:
    P60842
  • Purity:
    95 - 100%
  • Carrier-Free:
    Y
  • Source:
    E. coli
  • Nuclease-free:
    N
  • Protein/Peptide Type:
    Recombinant
  • Gene ID:
    1973
  • Formulation:
    Lyophilized from PBS, pH 7.4 containing 0.01% SKL, 5% trehalose
  • Protein/Peptide Name:
    EIF4A1
  • Tag sequence:
    MGHHHHHHSGSEF
  • Endotoxin-free:
    N
  • Species:
    Human
  • Protein Function:
    Bio-Markers & CD Antigens
  • Animal-Free:
    Y
  • Tested Applications:
    Positive control, Immunogen, SDS-PAGE, Western blot.
  • Endotoxin Content:
    <1.0 EU per 1 μg (determined by the LAL method)
  • Sequence:
    Met1~Thr271
  • Concentration:
    0.2 mg/ml
  • Cat. No.:
    MSPP-RPF009HU1
  • Shipping Temperature:
    4 °C
  • Protein Synonyms:
    Eukaryotic Translation Initiation Factor 4A1
  • Conjugation:
    Unconjugated

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.