Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant CAV1 (prokaryotic)(from E. coli)
Human Recombinant CAV1 (prokaryotic)(from E. coli)
Catalog # MSPP-RPA214HU1
Supplier:  Cloud-Clone
CAS Number:  
undefined
Human Recombinant CAV1 (prokaryotic)(from E. coli)
Catalog # MSPP-RPA214HU1
Supplier:  Cloud-Clone
Supplier Number:  RPA214HU01
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <1.0 EU per 1 μg (determined by the LAL method)
  • Gene ID:
    857
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Caveolin 1
  • UniProtKB:
    Q03135
  • Protein/Peptide Name:
    CAV1
  • Purity:
    97 - 100%
  • Molecular Weight:
    16 kDa
  • Sequence:
    Ser2~Ser104
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4 containing 0.01% SKL, 5% trehalose
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Positive control, Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-RPA214HU1

Specifications

About this item

This is a CAV1 recombinant protein (prokaryotic), Human is sequencing from Ser2~Ser104 with 97 to 100% purity. LyopH ilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

The scaffolding protein encoded by this gene is the main component of the caveolae plasma membranes found in most cell types. The protein links integrin subunits to the tyrosine kinase FYN, an initiating step in coupling integrins to the Ras-ERK pathway and promoting cell cycle progression. The gene is a tumor suppressor gene candidate and a negative regulator of the Ras-p42/44 MAP kinase cascade. CAV1 and CAV2 are located next to each other on chromosome 7 and express colocalizing proteins that form a stable hetero-oligomeric complex. By using alternative initiation codons in the same reading frame, two isoforms (alpha and beta) are encoded by a single transcript from this gene.

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use In clinical diagnostic procedures. Please proper stored each component based on the instruction.