ContactUsLinkComponent
Eluator™ Vacuum Elution Device, Promega
Eluator™ Vacuum Elution Device, Promega
Catalog # PAA1071
CAS Number:  
Eluator™ Vacuum Elution Device, Promega
Catalog # PAA1071
Supplier Number:  A1071
CAS Number:  
Specifications
  • Description:
    Eluator Vacuum Elution Device, 4 each
  • Size:
    4 each
  • Cat. No.:
    PAA1071
  • Supplier no.:
    A1071
Specifications
About this item
The Eluator Vacuum Elution Device is used with PureYield Purification Systems to elute purified DNA or RNA without the need for centrifugation.
  • Use with PureYield™ Purification Systems to Elute Without Centrifugation
  • Increased DNA or RNA recovery compared to centrifugation protocols
  • Faster purification
This simple, but elegant piece of equipment eliminates the need for centrifugation for elution of DNA when using the PureYield Midipreps or Maxiprep Plasmid DNA Purification Systems. The Eluator Vacuum Elution Device consists of two pieces, a blue base and a clear column assembly. The base interfaces with a Vacuum Manifold that contains Luer-Lok fittings, and holds a 1.5ml tube to capture eluted nucleic acids. The column assembly accepts PureYield Midiprep or Maxiprep columns. The Eluator Device not only saves time by eliminating the requirement for a centrifuge with a swinging bucket rotor, but also increases recovery of material compared to the centrifugation protocol.
The Eluator™ device eliminates the need for a centrifuge with a swinging bucket rotor for nucleic acid purification for faster, more efficient purification protocols.
The base connects to a vacuum manifold via Luer-lock fittings and accommodates a 1.5 mL tube to capture the eluted nucleic acids.
Ordering information: This device requires the use of a vacuum pump and vacuum manifold.