Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant CRN (prokaryotic) (from E.coli)
Human Recombinant CRN (prokaryotic) (from E.coli)
Catalog # MSPP-RPG316HU3
CAS Number:  
undefined
Human Recombinant CRN (prokaryotic) (from E.coli)
Catalog # MSPP-RPG316HU3
Supplier Number:  RPG316HU03
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    <1.0 EU per 1 µg (determined by the LAL method)
  • Gene ID:
    10699
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Corin
  • UniProtKB:
    Q9Y5Q5
  • Protein/Peptide Name:
    CRN (prokaryotic)
  • Purity:
    90 - 100%
  • Molecular Weight:
    53 kDa
  • Sequence:
    Arg134~Pro573
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Positive control, Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-RPG316HU3
  • Supplier no.:
    RPG316HU03

Specifications

About this item

This is a CRN recombinant protein (prokaryotic), Human is sequencing from Arg134~Pro573 with 90 to 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

Corin encodes a member of the type II transmembrane serine protease class of the trypsin superfamily. The encoded protein converts pro-atrial natriuretic peptide to biologically active atrial natriuretic peptide, a cardiac hormone that regulates blood volume and pressure. This protein may also function as a pro-brain-type natriuretic peptide convertase. The corin cDNA encodes a deduced 1,042-amino acid, type II transmembrane protein. At its N terminus, corin contains a short cytoplasmic domain and a transmembrane sequence. In the extracellular region of corin, there are 2 frizzled-like cysteine-rich motifs, 7 low density lipoprotein receptor repeats, a macrophage scavenger receptor-like domain, and a trypsin-like protease domain at the C terminus.

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.